missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TLE2 Full-length ORF (NP_003251.2, 1 a.a. - 743 a.a.) Recombinant Protein with GST-tag at N-terminal
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
10 ug
25 ug
Conditionnement:
10µg
25µg
Description
Sequence: MYPQGRHPTPLQSGQPFKFSILEICDRIKEEFQFLQAQYHSLKLECEKLASEKTEMQRHYVMYYEMSYGLNIEMHKQAEIVKRLSGICAQIIPFLTQEHQQQVLQAVERAKQVTVGELNSLIGQQLQPLSHHAPPVPLTPRPAGLVGGSATGLLALSGALAAQAQLAAAVKEDRAGVEAEGSRVERAPSRSASPSPPESLVEEERPSGPGGGGKQRADEKEPSGPYESDEDKSDYNLVVDEDQPSEPPSPATTPCGKVPICIPARRDLVDSPASLASSLGSPLPRAKELILNDLPASTPASKSCDSSPPQDASTPGPSSASHLCQLAAKPAPSTDSVALRSPLTLSSPFTTSFSLGSHSTLNGDLSVPSSYVSLHLSPQVSSSVVYGRSPVMAFESHPHLRGSSVSSSLPSIPGGKPAYSFHVSADGQMQPVPFPSDALVGAGIPRHARQLHTLAHGEVVCAVTISGSTQHVYTGGKGCVKVWDVGQPGAKTPVAQLDCLNRDNYIRSCKLLPDGRSLIVGGEASTLSIWDLAAPTPRIKAELTSSAPACYALAVSPDAKVCFSCCSDGNIVVWDLQNQTMVRQFQGHTDGASCIDISDYGTRLWTGGLDNTVRCWDLREGRQLQQHDFSSQIFSLGHCPNQDWLAVGMESSNVEILHVRKPEKYQLHLHESCVLSLKFASCGRWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISRNNKYIVTGSGDKKATVYEVVY
Spécification
Spécification
Numéro d’adhésion | NP_003251.2 |
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 7089 |
Poids moléculaire | 106.2kDa |
Nom | TLE2 (Human) Recombinant Protein (P01) |
Méthode de purification | Glutathione Sepharose 4 Fast Flow |
Test du contrôle qualité | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantité | 25 ug |
Immunogène | MYPQGRHPTPLQSGQPFKFSILEICDRIKEEFQFLQAQYHSLKLECEKLASEKTEMQRHYVMYYEMSYGLNIEMHKQAEIVKRLSGICAQIIPFLTQEHQQQVLQAVERAKQVTVGELNSLIGQQLQPLSHHAPPVPLTPRPAGLVGGSATGLLALSGALAAQAQLAAAVKEDRAGVEAEGSRVERAPSRSASPSPPESLVEEERPSGPGGGGKQRADEKEPSGPYESDEDKSDYNLVVDEDQPSEPPSPATTPCGKVPICIPARRDLVDSPASLASSLGSPLPRAKELILNDLPASTPASKSCDSSPPQDASTPGPSSASHLCQLAAKPAPSTDSVALRSPLTLSSPFTTSFSLGSHSTLNGDLSVPSSYVSLHLSPQVSSSVVYGRSPVMAFESHPHLRGSSVSSSLPSIPGGKPAYSFHVSADGQMQPVPFPSDALVGAGIPRHARQLHTLAHGEVVCAVTISGSTQHVYTGGKGCVKVWDVGQPGAKTPVAQLDCLNRDNYIRSCKLLPDGRSLIVGGEASTLSIWDLAAPTPRIKAELTSSAPACYALAVSPDAKVCFSCCSDGNIVVWDLQNQTMVRQFQGHTDGASCIDISDYGTRLWTGGLDNTVRCWDLREGRQLQQHDFSSQIFSLGHCPNQDWLAVGMESSNVEILHVRKPEKYQLHLHESCVLSLKFASCGRWFVSTGKDNLLNAWRTPYGASIFQSKESSSVLSCDISRNNKYIVTGSGDKKATVYEVVY |
Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Abnova™ Human TLE2 Full-length ORF (NP_003251.2, 1 a.a. - 743 a.a.) Recombinant Protein with GST-tag at N-terminal >
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu