missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human TNF-RII (soluble) Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 310-12-1MG

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17892333

  • 2965.00€ / 1mg
En stock
pour voir le stock disponible 

Description

Description

TNFR2 (tumor necrosis factor receptor superfamily member 1B) is a receptor with high affinity for TNFSF2/TNF-alpha. It has a much lower affinity for homotrimeric TNFSF1/lymphotoxin-alpha. The TRAF1/TRAF2 complex recruits the apoptotic suppressors BIRC2 and BIRC3 to TNFRSF1B/TNFR2. TNFR2 mediates most of the metabolic effects of TNF-alpha.
Spécification

Spécification

P20333
protein with no preservative
18.9
1 mg
-20°C
<1 EU/μg
Tnfrsf1b
Unconjugated
MAPEPGSTCR LREYYDQTAQ MCCSKCSPGQ HAKVFCTKTS DTVCDSCEDS TYTQLWNWVP ECLSCGSRCS SDQVETQACT REQNRICTCR PGWYCALSKQ EGCRLCAPLR KCRPGFGVAR PGTETSDVVC KPCAPGTFSN TTSSTDICRP HQICNVVAIP GNASMDAVCT STSP
≥ 98% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay, Western Blot
7133
Human TNF-RII (soluble)
E. coli
RUO
CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1 B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1 B; Tumor necrosis factor receptor superfamily member 1 b, membrane form; tumor necrosis factor receptor superfamily, member 1 B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2
Human
Recombinant
Lyophilized
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions