missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TNPO3 Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0670
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
TNPO3 Synthetic Peptide, PEP-0670, from Invitrogen.
This peptide corresponds to 18 amino acids near the amino terminus of human TNPO3.PEP-0670 can be used as a blocking peptide with polyclonal antibody PA5-20550.Spécification
23534 | |
purified | |
Human TNPO3 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9Y5L0 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TNPO3 | |
0.02% sodium azide | |
50 μg | |
PBS |