Learn More
Abnova™ Human TRIM36 Full-length ORF (NP_001017397.1, 1 a.a. - 60 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00055521-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene. [provided by RefSeq]
Sequence: MSESGEMSEFGYIMELIAKGKMPDWRRGYRCRQGCGKTTELATATDFSQTGNKSGKHFKTSpécification
NP_001017397.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HAPRIN/RBCC728/RNF98 | |
TRIM36 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55521 | |
TRIM36 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSESGEMSEFGYIMELIAKGKMPDWRRGYRCRQGCGKTTELATATDFSQTGNKSGKHFKT | |
RUO | |
TRIM36 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |