Learn More
ACROBiosystems Human VEGF R2 / KDR Protein, Strep Tag
MALS verified
Marca: ACROBiosystems KDR-H5280-1mg
Detalles adicionales : Poids : 0.00010kg
Descripción
Kinase insert domain receptor (KDR) is also known as CD309, FLK1, VEGFR, VEGFR2, and is one of the subtypes of VEGFR. VEGF receptors are receptors for vascular endothelial growth factor (VEGF). There are three main subtypes of VEGFR, numbered 1, 2 and 3. The VEGF receptors have an extracellular portion consisting of 7 immunoglobulin-like domains, a single transmembrane spanning region and an intracellular portion containing a split tyrosine-kinase domain. VEGF-A binds to VEGFR-1 (Flt-1) and VEGFR-2 (KDR/Flk-1). VEGFR-2 appears to mediate almost all of the known cellular responses to VEGF.The function of VEGFR-1 is less well defined, although it is thought to modulate VEGFR-2 signaling. Another function of VEGFR-1 may be to act as a dummy/decoy receptor, sequestering VEGF from VEGFR-2 binding (this appears to be particularly important during vasculogenesis in the embryo). In addition, VEGFR2 is able to interact with HIV-1 extracellular Tat protein upon VEGF activation, and seems to enhance angiogenesis in Kaposi's sarcoma lesions.
This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 86.3 kDa. The protein migrates as 120-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAI31823 | |
1 mg | |
Ala 20 - Glu 764 | |
KDR,CD309,FLK1,VEGFR,VEGFR2 | |
Unconjugated | |
Strep Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
86.3 kDa | |
Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823). | |
-20°C | |
VEGF R2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |