Learn More
Abnova™ Human ZIM2 Full-length ORF (NP_056178.3, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Spécification
Numéro d’adhésion | NP_056178.3 |
---|---|
À utiliser avec (application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formule | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Identification génétique (Entrez) | 23619 |
Poids moléculaire | 87.6kDa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16107552
|
Abnova™
H00023619-P01.25UG |
25 ug |
508.00€
25µg |
Expédition estimée: 20-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16197542
|
Abnova™
H00023619-P01.10UG |
10 ug |
335.00€
10µg |
Expédition estimée: 20-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5' exons and have a common promoter, and both genes are paternally expressed (PMID:15203203). Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Additional 5' alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene. [provided by RefSeq]
Sequence: MYQPEDDNNSDVTSDDDMTRNRRESSPPHSVHSFSGDRDWDRRGRSRDMEPRDRWSHTRNPRSRMPPRDLSLPVVAKTSFEMDREDDRDSRAYESRSQDAESYQNVVDLAEDRKPHNTIQDNMENYRKLLSLGFLAQDSVPAEKRNTEMLDNLPSAGSQFPDFKHLGTFLVFEELVTFEDVLVDFSPEELSSLSAAQRNLYREVMLENYRNLVSLGHQFSKPDIISRLEEEESYAMETDSRHTVICQGESHDDPLEPHQGNQEKLLTPITMNDPKTLTPERSYGSDEFERSSNLSKQSKDPLGKDPQEGTAPGICTSPQSASQENKHNRCEFCKRTFSTQVALRRHERIHTGKKPYECKQCAEAFYLMPHLNRHQKTHSGRKTSGCNEGRKPSVQCANLCERVRIHSQEDYFECFQCGKAFLQNVHLLQHLKAHEAARVLPPGLSHSKTYLIRYQRKHDYVGERACQCCDCGRVFSRNSYLIQHYRTHTQERPYQCQLCGKCFGRPSYLTQHYQLHSQEKTVECDHCSpécification
NP_056178.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
87.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MYQPEDDNNSDVTSDDDMTRNRRESSPPHSVHSFSGDRDWDRRGRSRDMEPRDRWSHTRNPRSRMPPRDLSLPVVAKTSFEMDREDDRDSRAYESRSQDAESYQNVVDLAEDRKPHNTIQDNMENYRKLLSLGFLAQDSVPAEKRNTEMLDNLPSAGSQFPDFKHLGTFLVFEELVTFEDVLVDFSPEELSSLSAAQRNLYREVMLENYRNLVSLGHQFSKPDIISRLEEEESYAMETDSRHTVICQGESHDDPLEPHQGNQEKLLTPITMNDPKTLTPERSYGSDEFERSSNLSKQSKDPLGKDPQEGTAPGICTSPQSASQENKHNRCEFCKRTFSTQVALRRHERIHTGKKPYECKQCAEAFYLMPHLNRHQKTHSGRKTSGCNEGRKPSVQCANLCERVRIHSQEDYFECFQCGKAFLQNVHLLQHLKAHEAARVLPPGLSHSKTYLIRYQRKHDYVGERACQCCDCGRVFSRNSYLIQHYRTHTQERPYQCQLCGKCFGRPSYLTQHYQLHSQEKTVECDHC | |
RUO | |
ZIM2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23619 | |
ZIM2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF656 | |
ZIM2 | |
Recombinant | |
wheat germ expression system |