Learn More
Proteintech™ HumanKine™ Recombinant Human GDNF Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Marque:  Proteintech™ HZ-1311-100UG
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16831426
  • 723.00€ / 100µg
Expédition estimée: 22-05-2024
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Spécification
2668
100 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Typically ≤ 10 ng/mL EC50
Lyophilized
33 to 36 kDa, homodimer,glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%