Learn More
Proteintech™ HumanKine™ Recombinant Human IFN Beta Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Marque:  Proteintech™ HZ-1298-1000UG
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16831416
  • 2701.00€ / 1000µg
Expédition estimée: 22-05-2024
pour voir le stock disponible 
Description
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Spécification
3456
1000 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water.
Typically ≤ 0.1 ng/mL EC50
Lyophilized
18 to 22 kDa, glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%