Learn More
Klebsiella aerogenes ATCC® 13048™†
For culture purposes and QC applications.
Marque: Microbiologics 0306P
Code nomenclature Nacres: NA.7
Informations supplémentaires : Poids : 0.50000kg Transport : UN number : 3373 Chem class : 6.2
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single strain of Klebsiella aerogenes in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2, 6 or 10
Spécification
Klebsiella aerogenes | |
1 | |
2°C to 8°C | |
Kwik-stik™ | |
Culture purpose and QC application |
ATCC 13048 | |
CE, online certificate of analysis | |
Lyophilized Pellet | |
KWIK-STIK | |
2/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.