Learn More
IgG Rabbit anti-Rabbit, Unconjugated, Polyclonal, Proteintech
Tous les produits Proteintech ProduitsDescription
The Rabbit IgG control antibody from Proteintech is a rabbit polyclonal antibody generated with n/a of Rabbit IgG control. This antibody recognizes rabbit antigen. The Rabbit IgG control antibody has been validated for the following applications: IP, WB, ELISA analysis.
Spécification
Spécification
| Antigène | IgG |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Espèces hôtes | Rabbit |
| Quantité | 1 mg |
| Primaire ou secondaire | Primary |
| Espèces cibles | Rabbit |
| Isotype | IgG |