Learn More
IL-10 Antibody (BT-063) [Janelia Fluor™ 585], Novus Biologicals™
Code produit 30169384
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30169384 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30169384 Fournisseur Novus Biologicals (Bio-Techne) Code fournisseur NBP328584JF585
Please to purchase this item. Need a web account? Register with us today!
Human Monoclonal Antibody
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spécification
Antigène IL-10
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone BT-063
Conjugué Janelia Fluor 585
Formule 50mM Sodium Borate
Alias de gène CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Espèces hôtes Human
Immunogène IL-10
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
Primaire ou secondaire Primary
Identification génétique (Entrez) 3586
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1