Learn More
Invitrogen™ IL-2 Monoclonal Antibody (S4B6)

Description
MM100 targets IL-2 in Western blot and ELISA applications and shows reactivity with mouse samples. The MM100 immunogen is recombinant mouse IL-2. MM100 detects IL-2 which has a predicted molecular weight of approximately 17 kDa. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
Spécification
Spécification
| Antigène | IL-2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | S4B6 |
| Concentration | 1.0 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with no preservative |
| Expression | IL2 |
| Numéro d’ordre du gène | P04351 |
| Alias de gène | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.