missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-5 Monoclonal Antibody (TRFK5)
GREENER_CHOICE

Code produit 11824341
Change view
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11824341 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11824341 Fournisseur Invitrogen™ Code fournisseur MM550C

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rat Monoclonal Antibody

MM550C targets IL-5 in ELISA, IA, and WB applications and shows reactivity with mouse samples. The MM550C immunogen is recombinant mouse IL-5. MM550C detects IL-5 which has a predicted molecular weight of approximately 13 kDa. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified. This product is a Low Endotoxin formulation. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY

Spécification

Antigène IL-5
Applications ELISA, Inhibition Assays, Western Blot
Classification Monoclonal
Clone TRFK5
Concentration 1.0 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative
Expression Il5
Numéro d’ordre du gène P04401
Alias de gène B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Symboles de gène(s) Il5
Espèces hôtes Rat
Immunogène Recombinant mouse IL-5.
Méthode de purification Protein A
Quantité 500 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 16191
Espèces cibles Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG1
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.