Learn More
Integrin alpha-5 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech
Code produit 16877033 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16877033 20 μL 20µL
16867033 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16877033 Fournisseur Proteintech Code fournisseur 105691AP20UL
Rabbit Polyclonal Antibody
Integrin alpha-5, also known as FNRA or VLA5A, is a protein that in humans is encoded by the ITGA5 gene. The product of this gene belongs to the integrin alpha chain family. Integrins are integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signaling.
Spécification
Antigène Integrin alpha-5
Applications Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Expression ITGA5
Numéro d’ordre du gène P08648, P11688
Alias de gène CD49e, FNRA, Integrin alpha 5, Integrin alpha F, ITGA5, VLA 5, VLA5A
Symboles de gène(s) ITGA5
Espèces hôtes Rabbit
Immunogène Integrin alpha-5 Fusion Protein Ag0860
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 16402, 3678
Espèces cibles Non-human Primate, Human, Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG