missing translation for 'onlineSavingsMsg'
Learn More

KLF7 Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene

Product Code. 17536324
Encompass_Preferred
Change view
Click to view available options
Quantité:
30 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
17536324 30 μL
17516324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17536324 Supplier Origene Technologies Supplier No. TA812008S

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF7 Monoclonal antibody specifically detects KLF7 in Human samples. It is validated for Western Blot

Specifications

Antigène KLF7
Applications Western Blot
Classification Monoclonal
Clone OTI1A7
Concentration 0.5 to 1.0 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Expression Klf7
Numéro d’ordre du gène O75840
Alias de gène 9830124P08Rik; Klf7; Krueppel-like factor 7; Kruppel like factor 7; Kruppel-like factor 7 (ubiquitous); Ubiquitous krueppel-like factor; ubiquitous Kruppel-like factor; ubiquitous Kruppel-like transcription factor; UKLF
Espèces hôtes Mouse
Immunogène Full length human recombinant protein of human KLF7 produced in E.coli.
Méthode de purification Affinity chromatography
Quantité 30 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 8609
Espèces cibles Human
Contenu et stockage -20° C, Avoid Freeze/Thaw Cycles
Forme Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.