missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KLPKKKNKHI PRPPQPGSPN YCKPDPPPPP PPHPHHGSGG ISPPPPITPP SMGVTVAGAY PAGPHTHPGL LRGGAGGLGI MGLPPLPAPG EPCPLAQEEV IEINRA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes.
Spécification
Spécification
| Antigène | KV3.3 (KCNC3) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Expression | KCNC3 |
| Numéro d’ordre du gène | Q14003 |
| Alias de gène | KCNC3; Kcr2-3; KShIIID; KV3.3; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily C member 3; potassium channel, voltage gated Shaw related subfamily C, member 3; potassium channel, voltage gated Shaw-related subfamily C, member 3; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; SCA13; Shaw type potassium channel Kv3.3; Shaw-related voltage-gated potassium channel protein 3; voltage-gated potassium channel protein KV3.3; voltage-gated potassium channel subunit Kv3.3 |
| Symboles de gène(s) | KCNC3 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?
