Learn More
Coverguard™ Gants en latex / coton
Click to view available options
Dimensions:
7
8
9
Conditionnement:
10 paires
Description
- PPE Category III
- Medium chemical protection
- Protect against bacteria and fungi
- Suitable for Forestry, Fishing, Horticulture, Farming, Waste treatment, Food-processing industries, Construction, Chemical
- Cotton inner for added comfort
- Blue natural latex coating
- Ideal for handling heavy objects in slippery and difficult environments
- Hangable individual pack
Spécification
Spécification
| Certifications / conformité (Europe) | EPI PPE Cat. III, EN 420:2003 + A1:2009, EN 388:2016, EN 374-1:2016, EN ISO 374-5:2016, Food contact |
| Applications | Industrial |
| Couleur | Bleu |
| Finger Thickness (English) | 51 mil |
| Finger Thickness (Metric) | 1.3 mm |
| Matériau | Latex, Cotton |
| Type de produit | Gants de protection chimique |
| Dimensions | 7 |
| Épaisseur (impériale) | 51 mil |
| Épaisseur (métrique) | 1.3 mm |
| Afficher plus |