missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMAN1 Mouse anti-Human, Clone: OTI2B10, lyophilized, TrueMAB™
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
Spécification
Spécification
| Antigène | LMAN1 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | OTI2B10 |
| Conjugué | Unconjugated |
| Formule | PBS with 8% trehalose and no preservative; pH 7.3 |
| Expression | LMAN1 |
| Numéro d’ordre du gène | P49257 |
| Alias de gène | ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58 |
| Symboles de gène(s) | LMAN1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?