Learn More
Thermo Scientific Chemicals Solutions oxydantes et de luminol (chaque flacon)
Code produit. p-7042139
Click to view available options
Quantité:
250 ml
500 ml
Conditionnement:
250mL
500mL
Code produit. 15474059
Marque: Thermo Scientific Chemicals J61462.AK
Demander un format en vrac ou personnalisé
Pour la détection de chimiluminécents des systèmes de transfert Western basés sur HRP
Used by biologists in cellular assays for the detection of copper, iron, and cyanides, as well as of specific proteins by western blot. For chemiluminecent HRP-based detection of western blotting systems. Microestimation of glucose and glucose oxidase using enzyme-induced chemiluminescence6,7 - Inhibitor of poly ADP Ribose Synthase8 - Demonstrations of the phenomenon of chemiluminescence, using different sensitizers to produce different colors of light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Luminol and oxidizing solutions (each bottle)
Informations sur la solubilité It is soluble in most polar organic solvents but is insoluble in water.
Quantité 250 ml
Sensibilité Light sensitive
Forme physique Liquide