missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mammaglobin B Polyclonal Antibody
GREENER_CHOICE

Code produit 16633425
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16633425 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16633425 Fournisseur Invitrogen™ Code fournisseur PA599173

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Mammaglobin B.

Secretoglobins (also called lipophilins or mammaglobins) are small secreted proteins of ndocrine-responsive organs and mucosal epithelia that form multimeric complexes and correlate with the development of various human cancers. Lipophilin A and Lipophilin B are orthologs of prostatein estramustine- binding protein), the major secretory glycoprotein of the rat ventral prostate gland. Lipophilin A, also designated LIPA, LPHA and secretoglobin, family 1D, member 1 (SCGB1D1), is a component of a heterodimeric molecule present in human tears. Lipophilin B, also designated LIPB, LPHB and secretoglobin, family 1D, member 2 (SCGB1D2), mRNA can be overexpressed in breast tumors and shows a high degree of correlation with the mRNA expression profile of mammaglobin. Histological detection in breast tissue of Mammaglobin A, also designated MGB1 and secretoglobin, family 2A, member 2 (SCGB2A2) and Mammaglobin B, also designated MGB2, Lipophilin C, LPHC, UGB3 and SCGB2A2, is a reliable diagnostic marker for breast tumors.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigène Mammaglobin B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Expression SCGB2A1
Numéro d’ordre du gène O75556
Alias de gène C3.1; Lacryglobin; LIPHC; lipophilin C; lipophilin-C; LPHC; LPNC; mammaglobin 2; mammaglobin B; mammaglobin-2; Mammaglobin-B; MGB2; prostatein; Prostatein peptide C3; Prostatic steroid-binding protein C3; Psbp1; Psbpc3; SCGB2A1; Scgb2a2; Secretoglobin family 2A member 1; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 1; UGB3
Symboles de gène(s) SCGB2A1
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human SCGB2A1(Accession O75556), corresponding to amino acid residues L3-D53.
Méthode de purification Affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 4246
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.