Learn More
Lovibond™ MD600 Colorimeter Replacement Reagents: COD
For use with MD600 multi-parameter colorimeter
Marque: Lovibond™ 2420725
Code nomenclature Nacres: NB.61
Informations supplémentaires : CAS : 7664-93-9 Poids : 1.50000kg
Description
- Parameter: COD
- Method: Dichromate /H2SO4
Spécification
US EPA | |
1 | |
Water quality testing | |
150 Tests | |
20°C | |
COD Tube Test Reagent |
Yellow to Brown | |
Liquid | |
MC500 multi-parameter colorimeter | |
0 to 150 mg/L | |
Dichromate/H2SO4 |