Learn More
MDM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16817504 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16817504 20 μL 20µL
16807504 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16817504 Fournisseur Proteintech Code fournisseur 175751AP20UL
Rabbit Polyclonal Antibody
This gene encodes a nuclear protein similar to the mouse double minute 1 protein. The mouse gene is located in double minute (DM) chromatin particles, is amplified in the mouse transformed 3T3 cell line, and the encoded protein is able to bind to p53. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification
Antigène MDM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression MDM1
Numéro d’ordre du gène Q5PQN4, Q8TC05, Q9D067
Alias de gène MDM1, Nuclear protein MDM1
Symboles de gène(s) Mdm1
Espèces hôtes Rabbit
Immunogène MDM1 Fusion Protein Ag10514
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 17245, 314859, 56890
Espèces cibles Rat, Mouse, Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG