Learn More
Microbiologics™ Candida albicans ATCC® 66027™†
For culture purposes and QC applications
Marque: Microbiologics™ 0981P
Code nomenclature Nacres: NA.7
Informations supplémentaires : Poids : 0.80000kg Transport : UN number : 3373 Chem class : 6.2
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single Candida albicans strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2 or 6
- ATCC™ 66027™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
Candida albicans | |
1 | |
2°C to 8°C | |
Kwik-stik™ | |
Culture purpose and QC application |
66027 | |
CE, online certificate of analysis | |
Lyophilized Pellet | |
QC Microorganism | |
2 U |