Learn More
Microbiologics™ X2 KWIK-STIK S. PSEUDINTERMEDIUS ATCC® 49444&asterisk;
For culture purposes and QC applications
Marque: Microbiologics™ 0907P
Code nomenclature Nacres: NA.7
Informations supplémentaires : CAS : 9000-70-8 Poids : 0.50000kg Transport : UN number : 3373 Chem class : 6.2
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single Staphylococcus pseudintermedius strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in pack of 2
- ATCC™ 49444™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
Staphylococcus aureus | |
2 | |
2°C to 8°C | |
Kwik-stik™ | |
Culture purpose and QC application |
49444 | |
CE, online certificate of analysis | |
Lyophilized Pellet | |
QC Microorganism | |
2 U |