Learn More
CCT2, Mouse, Clone: 5B5C4, Abnova™
Description
This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the gene described in this record have been observed but have not been thoroughly characterized. [provided by RefSeq
Spécification
Spécification
| Antigène | CCT2 |
| Applications | ELISA, Flow Cytometry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 5B5C4 |
| Conjugué | Unconjugated |
| Description | Mouse monoclonal antibody raised against partial recombinant CCT2. |
| Dilution | ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
| Formule | In ascites (0.03% sodium azide) |
| Expression | CCT2 |
| Alias de gène | 99D8.1/CCT-beta/CCTB/MGC142074/MGC142076/PRO1633/TCP-1-beta |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.