Learn More
PTK2, Mouse, Clone: 10H7E9, Abnova™
Description
This gene encodes a cytoplasmic protein tyrosine kinase which is found concentrated in the focal adhesions that form between cells growing in the presence of extracellular matrix constituents. The encoded protein is a member of the FAK subfamily of protein tyrosine kinases but lacks significant sequence similarity to kinases from other subfamilies. Activation of this gene may be an important early step in cell growth and intracellular signal transduction pathways triggered in response to certain neural peptides or to cell interactions with the extracellular matrix. At least four transcript variants encoding four different isoforms have been found for this gene, but the full-length natures of only two of them have been determined. [provided by RefSeq
Spécification
Spécification
| Antigène | PTK2 |
| Applications | ELISA, Flow Cytometry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 10H7E9 |
| Conjugué | Unconjugated |
| Description | Mouse monoclonal antibody raised against partial recombinant PTK2. |
| Dilution | ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
| Formule | In ascites (0.03% sodium azide) |
| Expression | PTK2 |
| Alias de gène | FADK/FAK/FAK1/pp125FAK |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.