Learn More
TF, Mouse, Monoclonal Antibody, Abnova™
Description
This gene encodes a glycoprotein with an approximate molecular weight of 76.5kDa. It is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds one ion of ferric iron. The function of this protein is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. This protein may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter and allergens from serum. [provided by RefSeq
Spécification
Spécification
| Antigène | TF |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Description | Mouse monoclonal antibody raised against native TF. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formule | In PBS (0.09% sodium azide) |
| Expression | TF |
| Alias de gène | DKFZp781D0156/PRO1557/PRO2086 |
| Symboles de gène(s) | TF |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.