Learn More
ACROBiosystems Mouse B7-H3 / CD276 Protein, Fc Tag
MALS verified
387.09€ - 2419.31€
Spécification
Numéro d’adhésion | NP_598744 |
---|---|
Poids moléculaire | 50.1 kDa |
Source | Mouse B7-H3, Fc Tag (B73-M5255) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Phe 244 (Accession # NP_598744). |
Immunogène | Val 29 - Phe 244 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15865957
|
ACROBiosystems
B73-M5255-100UG |
100 μg |
387.09€
100µg |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15875957
|
ACROBiosystems
B73-M5255-1MG |
1 mg |
2419.31€
1mg |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
B7 homolog 3 (B7-H3), a member of the immunoglobulin superfamily, is also known CD276, which contains two Ig-like C2-type (immunoglobulin-like) domains and two Ig-like V-type (immunoglobulin-like) domains. B7-H3 may participate in the regulation of T-cell-mediated immune response. B7-H3 also plays a protective role in tumor cells by inhibiting natural-killer mediated cell lysis as well as a role of marker for detection of neuroblastoma cells. Furthermore, B7-H3 is involved in the development of acute and chronic transplant rejection and in the regulation of lymphocytic activity at mucosal surfaces. It could also play a key role in providing the placenta and fetus with a suitable immunological environment throughout pregnancy.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_598744 | |
Mouse B7-H3, Fc Tag (B73-M5255) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Phe 244 (Accession # NP_598744). | |
-20°C | |
B7-H3 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
50.1 kDa | |
Val 29 - Phe 244 | |
B7-H3,CD276,B7 homolog 3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |