missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Mouse Glycophorin A / CD235a Protein, Fc,His Tag
350.80€ - 1463.68€
Spécification
Poids moléculaire | 39.1 kDa |
---|---|
Source | Mouse Glycophorin A, Fc,His Tag (CDA-M526x) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 108 (Accession # P14220). |
Immunogène | Met 1 - Val 108 |
Conditions de stockage | -20°C |
Alias de gène | CD235a,Glycophorin-A,Gypa |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15856357
|
ACROBiosystems
CDA-M526X-50UG |
50 μg |
350.80€
50µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15846357
|
ACROBiosystems
CDA-M526X-500UG |
500 μg |
1463.68€
500µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Glycophorin-A, a member of the glycophorin-A family, is also known as CD235a and Gypa, which is heavily glycosylated on serine and threonine residues at the N-terminal extracellular domain. CD235a,a single-pass type III membrane protein,is the major intrinsic membrane sialoglycoprotein of the erythrocyte. Also,CD235a appears to be important for the function of SLC4A1 and is required for high activity of SLC4A1. Furthermore,CD235a may be involved in translocation of SLC4A1 to the plasma membrane.
This protein carries a human IgG1 Fc tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
39.1 kDa | |
Met 1 - Val 108 | |
CD235a,Glycophorin-A,Gypa | |
Unconjugated | |
His Tag and Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Mouse Glycophorin A, Fc,His Tag (CDA-M526x) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 108 (Accession # P14220). | |
-20°C | |
Glycophorin A | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |