missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-10, Animal-Free Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ AF-210-10-500UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17893183

  • 3282.00€ / 500µg
Expédition estimée: 18-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.
Spécification

Spécification

P18893
protein with no preservative
18.7
500 µg
-20°C
<0.1 EU/µg
IL10
Unconjugated
MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGQKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLEDQGVYKA MNEFDIFINC IEAYMMIKMK S
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
16153
Mouse IL-10, Animal-Free
E. coli
RUO
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Mouse
Recombinant
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions