missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-17D Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 210-17D-100UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17890333

  • 577.00€ / 100µg
Expédition estimée: 12-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. The treatment of endothelial cells with this cytokine has been shown to stimulate the production of other cytokines including IL6, IL8 and CSF2/ GM-CSF. The increased expression of IL8 induced by this cytokine was found to be NF-kappa B-dependent.
Spécification

Spécification

ELISA, Functional Assay
239114
Mouse IL-17 D
E. coli
RUO
AI462269; FLJ30846; IL17D; IL-17 D; IL-22; IL-27; Il27a; IL-3 R; interleukin 17 D; interleukin 27 A; interleukin-17 D; Interleukin-27; UNQ3096/PRO21175
Mouse
Recombinant
Lyophilized
protein with no preservative
40.6
100 µg
-20°C
<1 EU/µg
IL17D
Unconjugated
AGALRTGRRP ARPRDCADRP EELLEQLYGR LAAGVLSAFH HTLQLGPREQ ARNASCPAGG RAADRRFRPP TNLRSVSPWA YRISYDPARF PRYLPEAYCL CRGCLTGLYG EEDFRFRSTP VFSPAVVLRR TAACAGGRSV YAEHYITIPV GCTCVPEPDK SADSANSSMD KLLLGPADRP AGR
≥ 95% by SDS-PAGE gel and HPLC analyses.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions