missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse Neuropoietin Recombinant Protein, PeproTech®

E. coli Recombinant Protein

Marque:  Gibco™ 250-25B-250UG

Informations supplémentaires : Poids : 0.16500kg

 Afficher plus de versions de ce produit

Code produit. 17891823

  • 1311.00€ / 250µg
Expédition estimée: 18-10-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Neuropoietin is a newly identified member of the IL-6 cytokine family. Members of this family, including IL-6, IL-11, oncostatin M, leukemia inhibitory factor (LIF), cardiotrophin-1 (CT-1), cardiotrophin-like cytokine, and CNTF, display a four-helix bundle structure, and signal through gp130-containing receptor complexes. Neuropoietin, which is predominantly expressed in neuroepithelia during embryonic life, acts through a receptor complex formed of a CNTF receptor-alpha component, gp 130, and LIF receptor. Like CNTF, it promotes the survival of embryonic motor neurons, and could increase the proliferation of neural precursor cells in the presence of EGF and FGF-2. Interestingly, the human neuropoietin gene has evolved toward a pseudogene, suggesting that alternative signaling via CNTF is an effective compensatory pathway.
Spécification

Spécification

P83714
protein with no preservative
19.8
250 µg
-20°C
<1 EU/µg
Ctf2
Unconjugated
MAPISPSEPI GQAYSLALYM QKNTSALLQT YLQHQGSPFS DPGFSAPELQ LSTLPSAAVS FKTWHAMEDA ERLSRAQGAF LALTQHLQLV GDDQSYLNPG SPILLAQLGA ARLRAQGLLG NMAAIMTALG LPIPPEEDTL GFVPFGASAF ERKCRGYIVT REYGHWTDRA VRDLALLKAK YSA
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
244218
Mouse Neuropoietin
E. coli
RUO
cardiotrophin 2; cardiotrophin-2; CT-2; Ctf2; CTF2P; Gm494; Neuropoietin; Np
Mouse
Recombinant
Lyophilized
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions