Learn More
Mouse Nogo Receptor/NgR Antibody, R&D Systems™
GREENER_CHOICE
Code produit 18756517
Change view
Click to view available options
Quantité:
100 μg
25 μg
Conditionnement:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18756517 25 μg 25µg
18011522 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18756517 Fournisseur R&D Systems Code fournisseur AF1440SP
Goat Polyclonal Antibody has been used in 12 publications
Nogo Receptor/NgR Polyclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Nogo Receptor/NgR
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugué Unconjugated
Dilution Western Blot 0.1 ug/mL
Formule Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Numéro d’ordre du gène Q99PI8
Alias de gène NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R
Symboles de gène(s) RTN4R
Espèces hôtes Goat
Immunogène Mouse myeloma cell line NS0-derived recombinant mouse NgR Cys27-Ser447 Accession # Q99PI8
Méthode de purification Affinity Purified
Quantité 25 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 65078
Spécificité du test Detects mouse Nogo Receptor in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human (rh) NgR is observed and less than 1% cross-reactivity with rhNgRH1 and rhNgRH2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Espèces cibles Mouse
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Afficher plus Afficher moins
For Research Use Only