missing translation for 'onlineSavingsMsg'
Learn More

Mouse/Rat CD31/PECAM-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Code produit p-4951330
Modifier l'affichage
Click to view available options
Quantité:
100 Tests
25 Tests
Conditionnement:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18741278 25 Tests 25 tests
18789107 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18741278 Fournisseur R&D Systems Code fournisseur FAB3628G025

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Goat Polyclonal Antibody has been used in 16 publications

CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Spécification

Antigène CD31/PECAM-1
Applications Flow Cytometry
Classification Polyclonal
Conjugué Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formule Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Numéro d’ordre du gène Q08481
Alias de gène adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Symboles de gène(s) PECAM1
Espèces hôtes Goat
Immunogène Mouse myeloma cell line NS0-derived recombinant mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481
Méthode de purification Affinity Purified
Quantité 25 Tests
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 5175
Spécificité du test Detects mouse CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD31 and recombinant porcine CD31 is observed. Detects mouse CD31 and rat CD31 in flow cytometry.
Espèces cibles Mouse, Rat
Contenu et stockage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Afficher plus Afficher moins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.