Learn More
STEMCELL Technologies Mouse Recombinant M-CSF Code produit.: 17129471
STEMCELL Technologies Mouse Recombinant M-CSF
Code produit. 17129471
10 μg
Code nomenclature Nacres: NA.28
Encompass_Preferred
Click to view available options
Quantité:
10 μg
100 μg
1 000 μg
Conditionnement:
1000µg
100µg
Code produit. 17129471
Marque: STEMCELL Technologies 78059.1
Macrophage colony-stimulating factor
  • Macrophage colony-stimulating factor (M-CSF) is a homodimeric glycoprotein growth factor that regulates proliferation and differentiation of myeloid hematopoietic progenitors to mononuclear phagocytic cell lineages, including monocytes, macrophages, and osteoclasts
  • M-CSF is a crucial factor for the development of tissue-resident macrophages in most tissues
  • It is required for the maturation and activation of monocytes and macrophages, and regulates inflammatory responses in conjunction with other stimuli such as IFN-γ, LPS, and IL-4
  • M-CSF is also required for bone resorption by osteoclasts, and is involved in the development and regulation of placenta, mammary gland, and brain
  • M-CSF is produced by monocytes, fibroblasts, osteoclasts, stromal cells, endothelial cells, and tumor cells
Spécification
Nom Colony stimulating factor 1, CSF-1
Quantité 10 μg
Type de cellule Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Monocytes; Myeloid Cells; Neurons
Pureté ou qualité ≥95%
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
STEMCELL Technologies Mouse Recombinant M-CSF