Learn More
STEMCELL Technologies Mouse Recombinant MIP-1 beta (CCL4)
Code nomenclature Nacres: NA.28
Click to view available options
Quantité:
10 μg
100 μg
Conditionnement:
10µg
Description
- Macrophage inflammatory protein-1 beta (MIP-1 beta), also known as CCL4, is a member of the CC family of chemokines and is most closely related to CCL3 (MIP-1 alpha)
- Cellular sources of MIP-1 beta include activated leukocytes (monocytes and T and B cells), brain endothelial cells, and smooth muscle cells
- MIP-1 beta, MIP-1 alpha, and RANTES have been shown to be major HIV-suppressive factors, possibly through the interactions of these chemokines with the receptor CCR5 on CD4+ T cells, which is also a major receptor for HIV entry into CD4+ T cells
- MIP-1 beta attracts a variety of immune cells to sites of microbial infection
- In addition to its chemotactic functions, MIP-1 beta induces the release of proinflammatory cytokines, mast cell degranulation, and NK cell activation
Spécification
Spécification
Nom | ACT-2, Immune activation protein 2, LAG-1, Lymphocyte activation gene 1 protein, MIP-1b, Protein H400, SCYA2, SCYA4, Small-inducible cytokine A4, T-cell activation protein 2 |
Quantité | 10 μg |
Type de cellule | B Cells; Dendritic Cells; Mesenchymal Stem and Progenitor Cells; Monocytes; NK Cells; Other; T Cells; T Cells, CD4+; T Cells, CD8+ |
Pureté ou qualité | ≥95% |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
STEMCELL Technologies Mouse Recombinant MIP-1 beta (CCL4)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu