missing translation for 'onlineSavingsMsg'
Learn More

Mouse TLR1 Antibody, R&D Systems™
GREENER_CHOICE

Artikelnummer. 18102721
Skift visning
Klik for at se tilgængelige muligheder
Quantité:
100 μg
25 μg
Pakningsstørrelse:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantité unitSize
18102721 100 μg 100µg
18781958 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18102721 Leverandør R&D Systems Leverandørnr. MAB1475

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Denne vare kan ikke returneres. Se returpolicy

Rat Monoclonal Antibody

TLR1 Monoclonal specifically detects TLR1 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigène TLR1
Applications Western Blot
Classification Monoclonal
Clone 285923
Concentration LYOPH
Conjugué Unconjugated
Dilution Western Blot 1 ug/mL
Formule Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Numéro d’ordre du gène Q9EPQ1
Alias de gène CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1
Symboles de gène(s) TLR1
Espèces hôtes Rat
Immunogène Mouse myeloma cell line NS0-derived recombinant mouse TLR1 Ser25-Asp581 Accession # Q9EPQ1
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 7096
Spécificité du test Detects mouse TLR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-1 RI, II, 3, 6, 9, recombinant human IL-1 R8, rmIL-18 R, rmIL-18 Rb, rmMD-1, rmRP105, rmSIGIRR, rmTLR2, 3, 4, or 6 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Espèces cibles Mouse
Contenu et stockage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forme Purified
Isotype IgG2a
Vis mere Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.