missing translation for 'onlineSavingsMsg'
Learn More

MRPL13 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Product Code. 16815814 Shop All Proteintech Products
Change view
Click to view available options
Quantité:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
16815814 20 μL 20µL
16805814 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16815814 Supplier Proteintech Supplier No. 162411AP20UL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.

Specifications

Antigène MRPL13
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression MRPL13
Numéro d’ordre du gène Q9BYD1, Q9D1P0
Alias de gène L13, L13A, L13mt, MRP L13, MRPL13, RPL13, RPML13
Symboles de gène(s) MRPL13
Espèces hôtes Rabbit
Immunogène MRPL13 Fusion Protein Ag9105
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 28998, 299938, 68537
Espèces cibles Mouse, Human, Rat, Non-human Primate
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.