missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MTA1 Polyclonal Antibody
GREENER_CHOICE

Code produit 16396974
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16396974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16396974 Fournisseur Invitrogen™ Code fournisseur PA578277

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Predicted reactivity: Mouse (97%), Rat (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.
TRUSTED_SUSTAINABILITY

Spécification

Antigène MTA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.01% thimerosal; pH 7
Expression MTA1
Numéro d’ordre du gène Q13330, Q62599, Q8K4B0
Alias de gène metastasis associated 1; metastasis associated gene 1 protein; metastasis-associated protein MTA1; MTA1; Zg29
Symboles de gène(s) MTA1
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the center region of human MTA1. The exact sequence is proprietary.
Méthode de purification Antigen Affinity Chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 116870, 64520, 9112
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.