missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYBPC1 Polyclonal Antibody
GREENER_CHOICE

Code produit 15712241 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15712241 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15712241 Fournisseur Invitrogen™ Code fournisseur PA554304

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: DWTLVETPPG EEQAKQNANS QLSILFIEKP QGGTVKVGED ITFIAKVKAE DLLRKPTIKW FKGKWMDLAS KAGKHLQLKE TFERHSRVYT FEMQIIKAKD NFAGNYRCEV TYKDKFDSCS FDLEVHESTG TTPN Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the slow-type isoform designated MYBPC1. Both the fast-type (MYBPC2) and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. Multiple isoforms of MYBPC1 are known to exist, and are present in varying amounts in different skeletal muscles. It is thought that the MYBPC1 slow subfamily may play important roles in the assembly and stabilization of sarcomeric M- and A-bands and regulate the contractile properties of the actomyosin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigène MYBPC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugué Unconjugated
Formule PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Expression MYBPC1
Numéro d’ordre du gène Q00872
Alias de gène 8030451F13Rik; C-protein, skeletal muscle slow isoform; LCCS4; muscle C-protein; MyBP-C slow; Mybpc1; MYBPCC; MYBPCS; myosin binding protein C, slow type; myosin binding protein C, slow-type; myosin-binding protein C, slow-type; skeletal muscle C-protein; slow MyBP-C; Slow-type C-protein
Symboles de gène(s) MYBPC1
Espèces hôtes Rabbit
Immunogène Recombinant protein corresponding to Human MYBPC1. Recombinant protein control fragment (Product #RP-90849).
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 4604
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.