Learn More
MYT1 Rabbit anti-Human, Polyclonal, Proteintech
Code produit 16844725 Tous les produits Proteintech Produits
Modifier l'affichage
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16844725 20 μL 20µL
16834725 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16844725 Fournisseur Proteintech Code fournisseur 262041AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
The protein encoded by this gene is a member of a family of neural specific, zinc finger-containing DNA-binding proteins. This kinase preferentially phosphorylates and inactivates cell division cycle 2 protein (CDC2), and thus negatively regulates cell cycle G2/M transition. This kinase is associated with the membrane throughout the cell cycle. Its activity is highly regulated during the cell cycle. Protein kinases AKT1/PKB and PLK (Polo-like kinase) have been shown to phosphorylate and regulate the activity of this kinase. Alternatively spliced transcript variants encoding distinct isoforms have been reported.The protein binds to the promoter regions of proteolipid proteins of the central nervous system and plays a role in the developing nervous system.
Spécification
Antigène MYT1
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Expression MYT1
Numéro d’ordre du gène Q01538
Alias de gène C20orf36, KIAA0835, KIAA1050, MTF1, myelin transcription factor 1, Myelin transcription factor I, MYT1, MYTI, PLPB1
Symboles de gène(s) MYT1
Espèces hôtes Rabbit
Immunogène MYT1 Fusion Protein Ag24580
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 4661
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG