missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NCF2 (Human) Recombinant Protein
Human NCF2 full-length ORF ( AAH01606.1, 1 a.a. - 526 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00004688-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MSLVEAISLWNEGVLAADKKDWKGALDAFSAVQDPHSRICFNIGCMYTILKNMTEAEKAFTRSINRDKHLAVAYFQRGMLYYQTEKYDLAIKDLKEALIQLRGNQLIDYKILGLQFKLFACEVLYNIAFMYAKKEEWKKAEEQLALATSMKSEPRHSKIDKAMECVWKQKLYEPVVIPVGRLFRPNERQVAQLAKKDYLGKATVVASVVDQDSFSGFAPLQPQAAEPPPRPKTPEIFRALEGEAHRVLFGFVPETKEELQVMPGNIVFVLKKGNDNWATVMFNGQKGLVPCNYLEPVELRIHPQQQPQEESSPQSDIPAPPSSKAPGRPQLSPGQKQKEEPKEVKLSVPMPYTLKVHYKYTVVMKTQPGLPYSQVRDMVSKKLELRLEQTKLSYRPRDSNELVPLSEDSMKDAWGQVKNYCLTLWCENTVGDQGFPDEPKESEKADANNQTTEPQLKKGSQVEALFSYEATQPEDLEFQEGDIILVLSKVNEEWLEGECKGKVGIFPKVFVEDCATTDLESTRREV
Spécification
AAH01606.1 | |
neutrophil cytosolic factor 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NCF2 | |
GST |
4688 | |
Wheat germ expression system | |
10 μg | |
FLJ93058, NCF-2, NOXA2, P67-PHOX, P67PHOX | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |