Learn More
Invitrogen™ NDUFB4 Recombinant Rabbit Monoclonal Antibody (24GB500)
Code produit 30276611
Change view
Click to view available options
Quantité:
20 μL
100 μL
Conditionnement:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30276611 20 μL 20µL
30275904 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30276611 Fournisseur Invitrogen™ Code fournisseur MA557104
Rabbit Recombinant Monoclonal Antibody
NDUFB4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY
Spécification
Antigène NDUFB4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24GB500
Concentration 1.2 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Expression NDUFB4
Numéro d’ordre du gène O95168, Q9CQC7
Alias de gène 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex
Symboles de gène(s) NDUFB4
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human NDUFB4 (aa 1-129).
Méthode de purification Affinity chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 288088, 4710, 68194
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG