missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS1 Polyclonal Antibody
GREENER_CHOICE

Code produit 13492327
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
13492327 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13492327 Fournisseur Invitrogen™ Code fournisseur PA528220

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse brain, rat brain. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (82%), Chicken (86%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene belongs to the complex I 75 kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Spécification

Antigène NDUFS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugué Unconjugated
Formule 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Expression NDUFS1
Numéro d’ordre du gène P28331, Q66HF1, Q91VD9
Alias de gène 5830412M15Rik; 9930026A05Rik; CI-75k; CI-75kD; complex I 75kDa subunit; complex I, mitochondrial respiratory chain, 75-kD subunit; complex I-75kD; MGC26839; mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit; NADH dehydrogenase (ubiquinone) Fe-S protein 1; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase); NADH:ubiquinone oxidoreductase core subunit S1; NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial; NDUFS1; NDUFS-1; OTTHUMP00000206345; PRO1304
Symboles de gène(s) NDUFS1
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the center region of human NDUFS1. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 227197, 301458, 4719
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.