missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS5 Polyclonal Antibody
GREENER_CHOICE

Code produit 13471647
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
13471647 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 13471647 Fournisseur Invitrogen™ Code fournisseur PA527542

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Raji. Predicted reactivity: Pig (82%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Located in the mitochondrial inner membrane, mitochondrial complex I is the first and largest enzyme in the electron transport chain of oxidative phosphorylation. By oxidizing NADH that is produced in the Krebs cycle, this complex utilizes the two electrons to reduce ubiquinone to ubiquinol, thereby initiating the passage of electrons to successive complexes and ultimately leading to the reduction of oxygen to water. Mitochondrial complex I consists of over 40 subunits and is of considerable clinical interest since defects in any of the subunits can lead to various myopathies and neuropathies. As a subunit of mitochondrial complex I, NDUFS5 (NADH dehydrogenase [ubiquinone] iron-sulfur protein 5), also designated NADH-ubiquinone oxidoreductase 15 kDa subunit, is a 106 amino acid protein that is suggested to not be involved in catalysis. NDUFS5 is expressed ubiquitously in human tissues, with a relative higher expression in human heart, skeletal muscle, liver, kidney and fetal heart.
TRUSTED_SUSTAINABILITY

Spécification

Antigène NDUFS5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Expression NDUFS5
Numéro d’ordre du gène O43920
Alias de gène AI256693; CI-15 kDa; CI15K; CI-15k; Complex I-15 kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 5; NADH dehydrogenase (ubiquinone) Fe-S protein 5, 15kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 5b, 15kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 5; NADH:ubiquinone oxidoreductase 15 kDa IP subunit; NADH:ubiquinone oxidoreductase subunit S5; NADH-ubiquinone oxidoreductase 15 kDa subunit; Ndufs5; Ndufs5b
Symboles de gène(s) NDUFS5
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the center region of human NDUFS5. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 362588, 4725
Espèces cibles Human, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.