missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide nitrilotriacétique, Honeywell Fluka™
Click to view available options
Quantité:
50 g
6 x 50 g
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Nom chimique ou matériau | Nitrilotriacetic acid |
Point d’éclair | 100°C (212°F) |
Formule linéaire | N(CH2COOH)3 |
Quantité | 6 x 50 g |
Numéro UN | NONH for all modes of transport |
Poids de la formule | 191.14 |
Pourcentage de pureté | ≥99% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'