Learn More
Novus Biologicals™ alpha Satellite Repeat Primer
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
0.2 mL
0.5 mL
Conditionnement:
0.20mL
0.50mL
Description
- Primer sets come premixed with forward and reverse primers at a concentration of 10μM
- Primers are compatible with all real-time PCR chemistries
- Each primer set has been tested and validated by chromatin immunoprecipitation (ChIP)
- Tm for primer set is between 53.4° to 54°c
- Amplicon size of primer set is 150 base pairs
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Spécification
Spécification
| Type de produit | alpha Satellite Repeat Primer |
| Contenu et stockage | Store at –20°C. Avoid Free/Thaw Cycles |
| Quantité | 0.2 mL |
| À utiliser avec (application) | Chromatin Immunoprecipitation |