Learn More
Novus Biologicals™ Luciferase (firefly) Protein
Code produit. 18789353 Tous les produits Bio Techne Produits
Click to view available options
Quantité:
0,1 mg
0,5 mg
Conditionnement:
0.10mg
0.50mg
Code produit. 18789353
Marque: Novus Biologicals™ NBP1483550.1mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-550 of Firefly Luciferase The Recombinant Firefly Luciferase Protein is derived from E. coli. The Recombinant Firefly Luciferase Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Spécification
Quantité 0,1 mg