Learn More
NRF1/nuclear respiratory factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16810224 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16810224 20 μL 20µL
16800224 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16810224 Fournisseur Proteintech Code fournisseur 124821AP20UL
Rabbit Polyclonal Antibody
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for ""nuclear factor (erythroid-derived 2)-like 1"" which has an official symbol of NFE2L1.
Spécification
Antigène NRF1/nuclear respiratory factor 1
Applications Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression NRF1
Numéro d’ordre du gène Q16656, Q62792, Q9WU00
Alias de gène ALPHA PAL, NRF 1, NRF1, NRF1/nuclear respiratory factor 1, nuclear respiratory factor 1
Symboles de gène(s) Nrf1
Espèces hôtes Rabbit
Immunogène NRF1/nuclear respiratory factor 1 Fusion Protein Ag3172
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 18181, 312195, 4899
Espèces cibles Human, Rat, Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG