missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hairy Cell Leukemia Monoclonal Antibody (DBA.44)

Description
Recommended positive controls: Tonsil.
Hairy Cell Leukemia (HCL) is a rare form of mature B-cell cancer characterized by the presence of abnormal B-cells known as 'hairy cells.' These cells have a distinct genetic profile that plays a significant role in the diagnosis and management of the disease. HCL includes variants such as the hairy cell leukemia variant (HCL-V) and splenic diffuse red pulp lymphoma (SDRPL), which are often detected through specific immunophenotyping and genetic analyses. One critical aspect of these analyses involves the use of flow cytometry to detect lambda light chain restriction among B-cells displaying bright CD11c positivity. Advances in genetic profiling using techniques like clonal analysis with consensus primer polymerase chain reaction (PCR) for heavy chain genes have further refined diagnostic precision, distinguishing HCL from its variants.
Spécification
Spécification
| Antigène | Hairy Cell Leukemia |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | DBA.44 |
| Concentration | Conc. Not Determined |
| Conjugué | Unconjugated |
| Formule | Tissue culture supernatant with 0.09% sodium azide |
| Expression | NSA2 |
| Numéro d’ordre du gène | O95478 |
| Alias de gène | 5730427N09Rik; CDK105; FLJ94393; Hairy cell leukemia protein 1; HCLG1; HCL-G1; HUSSY29; HUSSY-29; L-name related protein; L-name-related protein 42; LNR42; NSA2; NSA2 ribosome biogenesis homolog; NSA2 ribosome biogenesis homolog (S. cerevisiae); NSA2, ribosome biogenesis homolog; Nsa2p; ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1; Tinp1; Yr29; YR-29 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?